Please login to your online account to display your discounted pricing

Alfa Aesar™ Gentamicin sulfate, 20mg/mL in distilled water, sterile-filtered


Quantity 50mL

Manufacturer: alfa aesar™  J67488AD

 View more versions of this product

Catalog No. AAJ67488AD

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

CAS 1405-41-0
Chemical Name or Material Gentamicin sulfate
MDL Number MFCD00270181
Health Hazard 2 GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Health Hazard 1 Danger
Name Note 20mg/mL in distilled water, sterile-filtered
RTECS Number LY2625000
Storage Note 1 -30° to -10°C
Health Hazard 3 GHS P Statement
P285-P261-P280-P342+P311-P363-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 50mL
Physical Form Liquid

Specifications

Chemical Name or Material Gentamicin sulfate
CAS 1405-41-0
Name Note 20mg/mL in distilled water, sterile-filtered
MDL Number MFCD00270181
Accessories

Danger

GHS P Statement
P285-P261-P280-P342+P311-P363-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

-30° to -10°C

SDS & Safety Documents