Please login to your online account to display your discounted pricing

Genlantis™ Turbo Dicer Kits


Rapid 2-hour digestion of dsRNA in vitro

Manufacturer: genlantis  T520001

 View more versions of this product

Catalog No. T520001

  • / Each
 

Description & Specifications

Specifications

Quantity 50 Tests
Includes Recombinant Turbo™ Dicer™ Enzyme kit (50U), TurboScript™ T7 transcription kit (5 rxns), GeneSilencer™ siRNA transfection reagent kit (50 rxns), RNA purification column 1 and 2
Storage Requirements -20°C
Storage Conditions -20°C
For Use With (Application) Generating small interfering RNAs (siRNAs) from up to 5 different genes and for 50 transfections in 24-well plates
Quantity 50 Tests