Genlantis™ Turbo Dicer Kits
Rapid 2-hour digestion of dsRNA in vitro
$1,225.98 - $2,393.00
Specifications
Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Application) Includes Quantity Price Quantity  
Catalog Number Mfr. No. For Use With (Application) Includes Quantity Price Quantity  
T520001 Genlantis
T520001
Generating small interfering RNAs (siRNAs) from up to 5 different genes and for 50 transfections in 24-well plates Recombinant Turbo™ Dicer™ Enzyme kit (50U), TurboScript™ T7 transcription kit (5 rxns), GeneSilencer™ siRNA transfection reagent kit (50 rxns), RNA purification column 1 and 2 50 Tests Each for $2,393.00
T520002 Genlantis
T520002
Use to cleave more than 95% of dsRNA template into 22 base pairings siRNAs within 2 hours under optimized reaction conditions Recombinant Turbo™ Dicer™ Enzyme kit (0.5U/μL), Turbo™ Dicer™ Reaction Buffer, 50mM MgCl2 solution, 10mM ATP, Dicer™ stop solution, Nuclease-free W 50U Each for $1,225.98
Specifications
Description
  • High yield of siRNA pool for multiple reactions
  • Effective and specific gene silencing
  • Wide range of accessory products and reagents
  • Ideal for use with GeneSilencer™ siRNA transfection reagent
  • The Recombinant Human Turbo Dicer Enzyme Kit uses an ultra-active form of human recombinant dicer enzyme that can cleave more than 95% of dsRNA template into 22bp siRNAs within 2 hours under optimized reaction conditions
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
0.0