Please login to your online account to display your discounted pricing
Genlantis™ Turbo Dicer Kits
Rapid 2-hour digestion of dsRNA in vitro
Manufacturer: genlantis  T520002
 View more versions of this product
Catalog No. T520002
  • / Each
Description & Specifications
Specifications
For Use With (Application) Use to cleave more than 95% of dsRNA template into 22 base pairings siRNAs within 2 hours under optimized reaction conditions
Quantity 50U
Includes Recombinant Turbo™ Dicer™ Enzyme kit (0.5U/μL), Turbo™ Dicer™ Reaction Buffer, 50mM MgCl2 solution, 10mM ATP, Dicer™ stop solution, Nuclease-free W
Quantity 50U
Storage Requirements -20°C
Storage Conditions -20°C