Genlantis™ SoluLyse-M™ Mammalian Protein Extraction Reagent
Efficient extraction of soluble proteins from mammalian cells
Manufacturer:  Genlantis L300500
 View more versions of this product
Catalog No. L300500
Specifications
Specifications
Tris 1X 25mM, pH 7.4 and Sucrose Buffer 250mM
Mammalian Protein Extraction Reagent
Room Temperature
500mL
Reporter assays, immunoassays and Protein purification, proprietary formulation of nonionic detergent optimized for whole-cell Protein extraction from cultured cells
Documents