Please login to your online account to display your discounted pricing

Genlantis™ SoluLyse-M™ Mammalian Protein Extraction Reagent


Efficient extraction of soluble proteins from mammalian cells

Manufacturer: genlantis  L300125

 View more versions of this product

Catalog No. L300125

  • / Each
 

Description & Specifications

Specifications

Storage Requirements Room Temperature
Formulation Tris 1X 25mM, pH 7.4 and Sucrose Buffer 250mM
Quantity 125mL
Product Type Mammalian Protein Extraction Reagent
For Use With (Application) Reporter assays, immunoassays and Protein purification, proprietary formulation of nonionic detergent optimized for whole-cell Protein extraction from cultured cells