Please login to your online account to display your discounted pricing

Genlantis™ SoluLyse-M™ Mammalian Protein Extraction Reagent

Efficient extraction of soluble proteins from mammalian cells

$279.92 - $646.69

Specifications

Product Type Mammalian Protein Extraction Reagent
Formulation Tris 1X 25mM, pH 7.4 and Sucrose Buffer 250mM
For Use With (Application) Reporter assays, immunoassays and Protein purification, proprietary formulation of nonionic detergent optimized for whole-cell Protein extraction from cultured cells
Storage Requirements Room Temperature
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

L300125

 
genlantis
L300125
125mL Each for $279.92

L300500

 
genlantis
L300500
500mL Each for $646.69
Description & Specifications

Specifications

Product Type Mammalian Protein Extraction Reagent
Formulation Tris 1X 25mM, pH 7.4 and Sucrose Buffer 250mM
For Use With (Application) Reporter assays, immunoassays and Protein purification, proprietary formulation of nonionic detergent optimized for whole-cell Protein extraction from cultured cells
Storage Requirements Room Temperature

  • Produces high yields of non-denatured proteins
  • Simple and rapid protocols optimized for both adherent and suspension cells
  • Proprietary non-ionic detergent designed for whole-cell protein extraction from cultured cells
  • Compatible with downstream applications
  • Gently and rapidly dissolves cell membranes at low concentrations without denaturing proteins

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.