Please login to your online account to display your discounted pricing

Genlantis™ Rat Neural Stem Cell (RNSC) Growth and Differentiation Media


Serum-free media optimized with growth factors

Manufacturer: genlantis  SMRD813250

 View more versions of this product

Catalog No. SMRD813250

  • / Each
 

Description & Specifications

Specifications

Solution Type RNSC Differentiation Medium
Quantity 250mL
Sterile Yes
Storage Requirements 4°C
Sterile Yes
Quantity 250mL