Please login to your online account to display your discounted pricing
Genlantis™ Rat Neural Stem Cell (RNSC) Growth and Differentiation Media
Serum-free media optimized with growth factors
$430.16 - $466.29
Specifications
Quantity 250mL
Sterile Yes
Storage Requirements 4°C
View More Specs
Products
Catalog Number Mfr. No. Solution Type Price Quantity    
SMRD813250
genlantis
SMRD813250
RNSC Differentiation Medium Each for $430.16
SMR813250
genlantis
SMR813250
RNSC Growth Medium Each for $466.29
Description & Specifications
Specifications
Quantity 250mL
Sterile Yes
Storage Requirements 4°C