Please login to your online account to display your discounted pricing

Genlantis™ PrimaPure™ Rat Neural Stem Cells (RNSC)


Isolated from cerebral striatum and mesencephalon of E14 fetal mouse and E18 fetal rat brain

Manufacturer: genlantis  SR82020F

 View more versions of this product

Catalog No. SR82020F

  • / Each
 

Description & Specifications

Specifications

Storage 4°C
Cell Type RNSC, Fetal
Animal Rat
Quantity >500000 Cells
For Use With (Application) In-vitro research use only