Please login to your online account to display your discounted pricing

Genlantis™ PrimaPure™ Rat Neural Stem Cells (RNSC)


Isolated from cerebral striatum and mesencephalon of E14 fetal mouse and E18 fetal rat brain

Manufacturer: genlantis  SR82020AK

 View more versions of this product

Catalog No. SR82020AK

  • / Each
 

Description & Specifications

Specifications

Storage 4°C
For Use With (Application) In-vitro research use only
Animal Rat
Cell Type RNSC, Adult, Complete kit
Quantity >500000 Cells
Includes An ampoule of cryopreserved RNSC, 250mL of Rat neural stem cell growth medium, and two non-tissue culture treated 10cm plates