missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Genlantis™ PrimaPure™ Rat Neural Stem Cells (RNSC)
Isolated from cerebral striatum and mesencephalon of E14 fetal mouse and E18 fetal rat brain
$2,778.73 - $3,600.71
Specifications
Content And Storage | 4°C |
---|---|
Species | Rat |
For Use With (Application) | In-vitro research use only |
Quantity | >500000 Cells |
Catalog Number | Mfr. No. | Species | Cell Type | Includes | Quantity | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Species | Cell Type | Includes | Quantity | Price | Quantity | ||
SR82020A
|
Genlantis™
SR82020A |
Rat | Rat Neural Stem Cells (RNSC, Adult) | Cells only | >500000 Cells |
N/A
|
|
||
SR82020AK
|
Genlantis™
SR82020AK |
Rat | Rat Neural Stem Cells (RNSC, Adult) | An ampoule of cryopreserved RNSC, 250mL of Rat neural stem cell growth medium, and two non-tissue culture treated 10cm plates | >500000 Cells |
N/A
|
|
||
SR82020FK
|
Genlantis™
SR82020FK |
Rat | Rat Neural Stem Cells (RNSC, Fetal) | An ampoule of cryopreserved RNSC, 250mL of Rat neural stem cell growth medium, and two non-tissue culture treated 10cm plates | >500000 Cells |
N/A
|
|
||
SR82020F
|
Genlantis™
SR82020F |
Rat | Rat Neural Stem Cells (RNSC, Fetal) | Cells only | >500000 Cells |
N/A
|
|
||
Description
These multipotent cells can be induced to differentiate into neurons, astrocytes and oligodendrocytes. Neural stem cells are a source of donor tissue for prospective neural cell therapy research given the ability of neurosphere to retain multi-lineage potential and proliferative ability in vitro.
- Also available from adult rat brain
- Maintained in an undifferentiated proliferative state by culturing them as free-floating neurospheres in serum-free medium optimized with growth factors
- NSC are cryopreserved at second passage and can be cultured and propagated for one to two passages prior to induction of phenotypic differentiation
- Each lot functionally characterized
- Each lot tested negative for mycoplasma, bacteria, yeast, and fungi
Specifications
4°C | |
Rat | |
In-vitro research use only | |
>500000 Cells |