Please login to your online account to display your discounted pricing

Genlantis™ PrimaPure™ Rat Mesenchymal Stem Cells (RMSC)


Ideal for investigating pathways of pluripotent cell differentiation into bone, cartilage, fat, and neuronal cells

Manufacturer: genlantis  SR49205K

 View more versions of this product

Catalog No. SR49205K

  • / Each
 

Description & Specifications

Specifications

Storage 4°C
Includes An ampoule of cryopreserved cells, 500mL of mesenchymal stem cell growth medium, and a subculture reagent kit (including 100mL each of HEPES buffered Saline solution, Trypsin/EDTA, and Trypsin neutralizing solution)
For Use With (Application) Investigating pathways of pluripotent cell differentiation into bone, cartilage, fat, and neuronal cells
Cell Type RMSC Kit
Animal Rat
Quantity >500000 Cells