Please login to your online account to display your discounted pricing

Genlantis™ PrimaPure™ Rat Mesenchymal Stem Cells (RMSC)


Ideal for investigating pathways of pluripotent cell differentiation into bone, cartilage, fat, and neuronal cells

Manufacturer: genlantis  SR49205

 View more versions of this product

Catalog No. SR49205

  • / Each
 

Description & Specifications

Specifications

Storage 4°C
For Use With (Application) Investigating pathways of pluripotent cell differentiation into bone, cartilage, fat, and neuronal cells
Includes Cells only
Animal Rat
Cell Type RMSC
Quantity >500000 Cells