Please login to your online account to display your discounted pricing

Genlantis™ PrimaPure™ Rat Mesenchymal Stem Cells (RMSC)

Ideal for investigating pathways of pluripotent cell differentiation into bone, cartilage, fat, and neuronal cells

$2,082.51 - $2,731.53

Specifications

Animal Rat
For Use With (Application) Investigating pathways of pluripotent cell differentiation into bone, cartilage, fat, and neuronal cells
Quantity >500000 Cells
Storage 4°C
View More Specs
Includes:  Complete System contains an ampule of cryopreserved cells (>500,000 cells), 500mL of mesenchymal stem cell growth medium, and a Subculture Reagent Kit (including 100mL each of HEPES Buffered Saline Solution, Trypsin/EDTA, and Trypsin Neutralizing Solution).
Products
Catalog Number Mfr. No. Includes Cell Type Price Quantity    

SR49205

 
genlantis
SR49205
Cells only RMSC Each for $2,082.51

SR49205K

 
genlantis
SR49205K
An ampoule of cryopreserved cells, 500mL of mesenchymal stem cell growth medium, and a subculture reagent kit (including 100mL each of HEPES buffered Saline solution, Trypsin/EDTA, and Trypsin neutralizing solution) RMSC Kit Each for $2,731.53
Description & Specifications

Specifications

Animal Rat
For Use With (Application) Investigating pathways of pluripotent cell differentiation into bone, cartilage, fat, and neuronal cells
Quantity >500000 Cells
Storage 4°C

Mesenchymal stem cells (MSC) have the ability to self-renew and differentiate into cells of various connective tissue lineages, such as bone, cartilage, fat, and neurons. The potential of MSCs to maintain multipotency and proliferate extensively in vitro provides new avenues for cell-based therapy in the restoration of damaged or diseased tissue.

  • Each lot is tested for its ability to attach and spread on standard tissue culture grade plastic surfaces and proliferate in Mesenchymal Stem Cell Growth Medium
  • Each lot is characterized for bone mineralization and lipid accumulation following differentiation using osteoblast and adipocyte differentiation media, repectively
  • Each lot tested negative for mycoplasma, bacteria, yeast, and fungi
  • 10 population doublings guaranteed with Genlantis media and reagents
  • Isolated from adult or fetal bone marrow
  • Cryopreserved at second passage