Please login to your online account to display your discounted pricing

Genlantis™ PrimaPure™ Human Umbilical Vein Endothelial Cells (HUVEC) and Kits

Functionally characterized

$898.84 - $1,531.18

Specifications

Cell Type Human Umbilical Vein Endothelial Cells (Neonatal)
Source Human Umbilical Vein
For Use With (Application) Used for physiological and pharmacological investigations, such as macromolecule transport, blood coagulation, fibrinolysis
Media Type Endothelial Cell Growth Medium
Sufficient For 16 Population Doublings
View More Specs
Includes:  Each kit contains an ampule of cryopreserved HUVEC (PH20005N), 500mL of Endothelial Cell Growth Medium (PM211500), and a Subculture Reagent Kit (PR090100K).
Products
Catalog Number Mfr. No. Quantity Price Quantity    

PH20005N

 
genlantis
PH20005N
>500000 Cells Each for $898.84

PH20005NP

 
genlantis
PH20005NP
Pooled, >500000 Cells Each for $898.84

PH20005NK

 
genlantis
PH20005NK
1 Kit Each for $1,531.18
Description & Specifications

Specifications

Cell Type Human Umbilical Vein Endothelial Cells (Neonatal)
Source Human Umbilical Vein
For Use With (Application) Used for physiological and pharmacological investigations, such as macromolecule transport, blood coagulation, fibrinolysis
Media Type Endothelial Cell Growth Medium
Sufficient For 16 Population Doublings

Human Umbilical Vein Endothelial Cells (HUVEC) are isolated from normal human umbilical vein. They are cryopreserved at the end of primary culture and can be cultured and propagated at least 16 population doublings. HUVEC are responsive to cytokine stimulation in the expression of cell adhesion molecules. These cell systems are commonly used for physiological and pharmacological investigations, such as macromolecule transport, blood coagulation and fibrinolysis.

  • Single donors to allow variability comparisons
  • Each lot tested negative for HIV, Hepatitis B and C, mycoplasma, bacteria, yeast, and fungi
  • Performance guaranteed with Genlantis media and reagents