Please login to your online account to display your discounted pricing

Genlantis™ PrimaPure™ Human Endothelial Cell Growth Media


Optimized for endothelial cells

Manufacturer: genlantis  PM114500

 View more versions of this product

Catalog No. PM114500

  • / Each
 

Description & Specifications

Specifications

Without Additives Serum, Growth Factor
Product Type Serum-Free; Growth Factor-Free
Storage Requirements 4°C
Without Additives Serum, Growth Factor
Quantity 500mL
Quantity 500mL