Genlantis™ PrimaPure™ Human Bronchial/Tracheal Epithelial Cells Growth Medium
Used to study the regulation of intracellular pH, the stimulant effect of IL-1b on airway epithelial cell growth, and the effect of human rhinovirus infection in terms of up regulation of ICAM-1 expression on epithelial cells
Supplier: Genlantis™ PM511GS
Description
Human Bronchial/Tracheal Epithelial Cells (HBEpC/HTEpC) are derived from the surface epithelium of normal human bronchi. These are cryopreserved at first passage and can be cultured and propagated at least 16 population doublings. Airway epithelial cells are targets in asthma, viral infection and fibrotic lung disease.
Specifications
4°C | |
Growth Medium | |
4 Vials |