Please login to your online account to display your discounted pricing
Genlantis™ PrimaPure™ Freezing Medium
Manufacturer: genlantis  PR04050E
 View more versions of this product
Catalog No. PR04050E
  • / Each
Description & Specifications
Specifications
Solution Type Endothelial Cell Freezing Medium
Quantity 50mL
Storage Requirements 4°C