Please login to your online account to display your discounted pricing

Genlantis™ Porcine Smooth Muscle Cells


Economical alternative to human smooth muscle cells

Manufacturer: genlantis  PP35205

 View more versions of this product

Catalog No. PP35205

  • / Each
 

Description & Specifications

Specifications

Storage 4°C
Animal Porcine
Cell Type PPASMC
For Use With (Application) Investigations in cardiac disease and arteriosclerosis, especially in co-culture with species-matched porcine arterial endothelial cells
Quantity >500000 Cells