Please login to your online account to display your discounted pricing
Genlantis™ Porcine Smooth Muscle Cells
Economical alternative to human smooth muscle cells
$1,790.76 - $1,790.76
Specifications
Animal Porcine
For Use With (Application) Investigations in cardiac disease and arteriosclerosis, especially in co-culture with species-matched porcine arterial endothelial cells
Quantity >500000 Cells
Storage 4°C
View More Specs
Products
Catalog Number Mfr. No. Cell Type Price Quantity    
PP35005
genlantis
PP35005
PCASMC Each for $1,790.76
PP35205
genlantis
PP35205
PPASMC Each for $1,790.76
PP35405
genlantis
PP35405
PAOSMC Each for $1,790.76
Description & Specifications
Specifications
Animal Porcine
For Use With (Application) Investigations in cardiac disease and arteriosclerosis, especially in co-culture with species-matched porcine arterial endothelial cells
Quantity >500000 Cells
Storage 4°C
  • Each lot functionally characterized
  • Same quality standard as the PrimaPure™ human cell products
  • Each lot tested negative for mycoplasma, bacteria, yeast, and fungi
  • Supports 16 population doublings (14 for PCASMC) with Genlantis™ media and reagents