Please login to your online account to display your discounted pricing

Genlantis™ Porcine Smooth Muscle Cell Kits


Supports 16 population doublings

Manufacturer: genlantis  PP35005K

 View more versions of this product

Catalog No. PP35005K

  • / Each
 

Description & Specifications

Specifications

Storage 4°C
Cell Type PCASMC
For Use With (Application) Investigations in cardiac disease and arteriosclerosis, especially in co-culture with species-matched porcine arterial endothelial cells
Animal Porcine
Quantity >500000 Cells
Includes Cryopreserved cells, growth medium, subculture reagent kit