Genlantis™ Porcine Smooth Muscle Cell Kits
Supports 16 population doublings
$2,389.35 - $2,389.35
Specifications
Animal Porcine
For Use With (Application) Investigations in cardiac disease and arteriosclerosis, especially in co-culture with species-matched porcine arterial endothelial cells
Includes Cryopreserved cells, growth medium, subculture reagent kit
Quantity >500000 Cells
Storage 4°C
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Cell Type Price Quantity  
Catalog Number Mfr. No. Cell Type Price Quantity  
PP35205K Genlantis
PP35205K
PPASMC Each for $2,389.35
PP35005K Genlantis
PP35005K
PCASMC Each for $2,389.35
PP35405K Genlantis
PP35405K
PAOSMC Each for $2,389.35
Specifications Description & Specifications
Specifications
Porcine
Cryopreserved cells, growth medium, subculture reagent kit
4°C
Investigations in cardiac disease and arteriosclerosis, especially in co-culture with species-matched porcine arterial endothelial cells
>500000 Cells