Genlantis™ Porcine Endothelial Cells
Economical alternative to human endothelial cells
Supplier: Genlantis™ PP30405
Description
- Each lot functionally characterized
- Same quality standard as the PrimaPure™ human cell products
- Each lot tested negative for mycoplasma, bacteria, yeast, and fungi
- Supports 16 population doublings (12 for PCAEC) with Genlantis™ media and reagents
Specifications
4°C | |
Porcine | |
>500000 Cells |
Porcine Aortic Endothelial Cells (PAOEC) | |
To study endothelial function and endothelial metabolism, especially in co-culture of species matched porcine arterial smooth muscle cells |