Please login to your online account to display your discounted pricing

Genlantis™ Porcine Endothelial Cells


Economical alternative to human endothelial cells

Manufacturer: genlantis  PP30205

 View more versions of this product

Catalog No. PP30205

  • / Each
 

Description & Specifications

Specifications

Storage 4°C
For Use With (Application) To study endothelial function and endothelial metabolism, especially in co-culture of species matched porcine arterial smooth muscle cells
Cell Type PPAEC
Animal Porcine
Quantity >500000 Cells