Genlantis™ Porcine Endothelial Cell Kits
Supports at least 16 population doublings
Supplier: Genlantis™ PP30405K
Specifications
4°C | |
Porcine | |
Cryopreserved cells, growth medium, subculture reagent kit |
Porcine Aortic Endothelial Cells (PAOEC) | |
To study endothelial function and endothelial metabolism, especially in co-culture of species matched porcine arterial smooth muscle cells | |
>500000 Cells |