missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Genlantis™ Porcine Endothelial Cell Kits
Supports at least 16 population doublings
$2786.00 - $2906.02
Specifications
Content And Storage | 4°C |
---|---|
Species | Porcine |
For Use With (Application) | To study endothelial function and endothelial metabolism, especially in co-culture of species matched porcine arterial smooth muscle cells |
Includes | Cryopreserved cells, growth medium, subculture reagent kit |
Quantity | >500000 Cells |
Catalog Number | Mfr. No. | Cell Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Cell Type | Price | Quantity | |||||
PP30005K
|
Genlantis™
PP30005K |
Porcine Coronary Artery Endothelial Cells (PCAEC) |
Each for $2,906.02
|
|
|||||
PP30405K
|
Genlantis™
PP30405K |
Porcine Aortic Endothelial Cells (PAOEC) |
Each for $2,786.00
|
|
|||||
PP30205K
|
Genlantis™
PP30205K |
Porcine Pulmonary Artery Endothelial Cells (PPAEC) |
Each for $2,786.00
|
|
|||||
Specifications
4°C | |
To study endothelial function and endothelial metabolism, especially in co-culture of species matched porcine arterial smooth muscle cells | |
>500000 Cells |
Porcine | |
Cryopreserved cells, growth medium, subculture reagent kit |