Please login to your online account to display your discounted pricing

Genlantis™ phCMV Vector Kits


Mammalian expression vector with significantly higher expression levels than traditional CMV promoter-based constitutive vectors

Manufacturer: genlantis  P003300

 View more versions of this product

Catalog No. P003300

  • / Each
 

Description & Specifications

Specifications

Promoter hCMV
Vector 25μg Supercoiled phCMV3 vector (1μg/mL), 10μg phCMV3/CAT positive control vector (1μg/mL)
Storage Requirements -20°C
Protein Expression Level High
Quantity 25μg
For Use With (Application) Encoding recombinant Proteins with C-terminal HA fusion tag, suitable for both stable (G418) and transient expression