Please login to your online account to display your discounted pricing

Genlantis™ phCMV Vector Kits


Mammalian expression vector with significantly higher expression levels than traditional CMV promoter-based constitutive vectors

Manufacturer: genlantis  P003200

 View more versions of this product

Catalog No. P003200

  • / Each
 

Description & Specifications

Specifications

Protein Expression Level High
Vector 25μg Supercoiled phCMV2 vector (1μg/mL), 10μg phCMV2/CAT positive control vector (1μg/mL)
For Use With (Application) Encoding recombinant Proteins with N-terminal HA fusion tag, suitable for both stable (G418) and transient expression
Quantity 25μg
Promoter hCMV
Storage Requirements -20°C
Size 25μg