Please login to your online account to display your discounted pricing
Genlantis™ phCMV Vector Kits
Mammalian expression vector with significantly higher expression levels than traditional CMV promoter-based constitutive vectors
Manufacturer: genlantis  P003100
 View more versions of this product
Catalog No. P003100
  • / Each
Description & Specifications
Specifications
Protein Expression Level High
Storage Requirements -20°C
Promoter hCMV
Quantity 25μg
For Use With (Application) Encoding recombinant Proteins in their native form, without any tags, suitable for both stable (G418) and transient expression
Vector 25μg Supercoiled phCMV1 vector (1μg/mL), 10μg phCMV1/CAT positive control vector (1μg/mL)