Genlantis™ phCMV Vector Kits
Mammalian expression vector with significantly higher expression levels than traditional CMV promoter-based constitutive vectors
$788.10 - $788.10
Specifications
Promoter hCMV
Protein Expression Level High
Quantity 25μg
Storage Requirements -20°C
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Application) Vector Price Quantity  
Catalog Number Mfr. No. For Use With (Application) Vector Price Quantity  
P003200 Genlantis
P003200
Encoding recombinant Proteins with N-terminal HA fusion tag, suitable for both stable (G418) and transient expression 25μg Supercoiled phCMV2 vector (1μg/mL), 10μg phCMV2/CAT positive control vector (1μg/mL) Each for $788.10
P003100 Genlantis
P003100
Encoding recombinant Proteins in their native form, without any tags, suitable for both stable (G418) and transient expression 25μg Supercoiled phCMV1 vector (1μg/mL), 10μg phCMV1/CAT positive control vector (1μg/mL) Each for $788.10
P003300 Genlantis
P003300
Encoding recombinant Proteins with C-terminal HA fusion tag, suitable for both stable (G418) and transient expression 25μg Supercoiled phCMV3 vector (1μg/mL), 10μg phCMV3/CAT positive control vector (1μg/mL) Each for $788.10
Specifications
Description
  • High-level constitutive transgene expression
  • Suitable for both stable (G418) and transient expression
  • Expression of native and HA-tagged proteins
  • Small size for increased transfection efficiency
  • Convenient multiple cloning site containing the most commonly-used restriction sites
  • Produce significantly higher expression levels than traditional human cytomegalovirus (CMV) promoter based constitutive expression vectors such as pcDNA3.1
Description & Specifications
Specifications
hCMV
High
25μg
-20°C