Please login to your online account to display your discounted pricing

Genlantis™ phCMV Reporter Vectors


High-level expression vectors for GFP or Luciferase fusion protein expression

Manufacturer: genlantis  P013500

 View more versions of this product

Catalog No. P013500

  • / Each
 

Description & Specifications

Specifications

Promoter hCMV
Marker Gene Kanamycin/Neomycin
Concentration 0.5μg/μL
Vector phCMV N-Luciferase vector
Protein Expression Level High
Reporter Gene Luciferase
For Use With (Application) Used for either transient transfection or for generating stable cells lines expressing the reporter gene alone or in fusion with gene of interest
Quantity 20μg
Storage Requirements -20°C
Size 20μg