Genlantis™ PerFectin™ Transfection Reagent
Optimized to provide efficient transfection of several popularly-used cell lines
$378.25 - $676.50
Specifications
For Use With (Application) | Efficient transfection of several popularly-used cell lines |
---|---|
Includes | 1 Vial PerFectin™ Lipid film, 1 x 0.8mL Hydration Buffer, 1 x 5.5mL DNA diluents |
Description
- High efficiency with CHO, COS, 293, HeLa, NIH-3T3, and Jurkat cells
- Easy and optimized protocols for individual cell lines
- Compatible with serum containing media
- Economical price
- Cationic lipid specifically formulated and optimized to provide efficient transfection of several popularly-used cell lines
Specifications
Efficient transfection of several popularly-used cell lines | |
1 Vial PerFectin™ Lipid film, 1 x 0.8mL Hydration Buffer, 1 x 5.5mL DNA diluents |