Please login to your online account to display your discounted pricing

Genlantis™ MycoScope™ PCR Detection Kit


Designed for sensitive detection of mycoplasma using PCR

Manufacturer: genlantis  MY01100

 View more versions of this product

Catalog No. MY01100

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Detect the presence of mycoplasma contamination in cultured cells
Includes 3 x 1.4mL dNTP/Buffer mixture (1X), 950μL primer mix (5X), 100μL positive control reaction tube, 1.5mL molecular biology grade water
No. of Reactions 100.0
Storage Requirements -20°C