missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Genlantis™ MycoScope™ PCR Detection Kit

Designed for sensitive detection of mycoplasma using PCR

$593.08 - $1,070.63

Specifications

For Use With (Application) Detect the presence of mycoplasma contamination in cultured cells
Storage Requirements -20°C
View More Specs

Includes: dNTP/Buffer mixture (1X); Primer mix (5X); Positive control reaction tube; Molecular biology grade water

Products 2
Catalog Number Mfr. No. Includes No. of Reactions Price Quantity  
Catalog Number Mfr. No. Includes No. of Reactions Price Quantity  
MY01050
View Documents Promotion Details
Genlantis™
MY01050
2 x 1.25mL dNTP/Buffer mixture 1X, 530μL primer mix 5X, 50μL positive control reaction tube, 1.5mL molecular biology grade water 50
Each for $593.08
 
MY01100
View Documents Promotion Details
Genlantis™
MY01100
3 x 1.4mL dNTP/Buffer mixture (1X), 950μL primer mix (5X), 100μL positive control reaction tube, 1.5mL molecular biology grade water 100
Each for $1,070.63
 
Description

Description

  • Allows detection of less than five mycoplasma genomes/μL sample
  • Unique primer sets detect all mycoplasma species commonly encountered in cell culture
  • Fast and easy protocol eliminate the need for labeling probes

Specifications

Specifications

Detect the presence of mycoplasma contamination in cultured cells
-20°C
SDS
Product Certifications