Please login to your online account to display your discounted pricing

Genlantis™ Mouse Neural Stem Cell (MNSC) Growth and Differentiation Medium


Serum-free media optimized with growth factors

Manufacturer: genlantis  SMM813250

 View more versions of this product

Catalog No. SMM813250

  • / Each
 

Description & Specifications

Specifications

Solution Type MNSC Growth Media
Quantity 250mL
Sterile Yes
Storage Requirements 4°C
Sterile Yes
Quantity 250mL