missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Genlantis™ Mouse Neural Stem Cell (MNSC) Growth and Differentiation Medium
Serum-free media optimized with growth factors
$323.71 - $552.83
Specifications
Content And Storage | 4°C |
---|---|
Product Type | Stem Cell Differentiation Media |
Sterility | Sterile |
Quantity | 250 mL |
Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
SMMD813250
|
Genlantis™
SMMD813250 |
Differentiation Media |
N/A
|
|
|||||
SMM813250
|
Genlantis™
SMM813250 |
Growth Media |
N/A
|
|
|||||
Specifications
4°C | |
Stem Cell Differentiation Media | |
Sterile | |
250 mL |