Please login to your online account to display your discounted pricing

Genlantis™ ImmunoPure™ HD Normal Human Peripheral Blood Mononuclear Cells (PBMNCs)


Consists of a variety of immune cells derived from the peripheral blood of normal adults, thereby qualifying them for a diverse number of cell-based experiments

Manufacturer: genlantis  PBMC002

 View more versions of this product

Catalog No. PBMC002

  • / Each
 

Description & Specifications

Specifications

Type Normal Human Peripheral Blood Mononuclear Cells
Certifications/Compliance IRB, FDA
Source Peripheral Blood of Normal Adults
Storage Requirements -170°C
For Use With (Application) Carry the duty of fighting infections as part of the immune system response
Media Type RPMI1640 medium + 10% (v/v) FBS, 2mM Glutamine, 1% (v/v) non-essential Amino Acids, 1% (v/v) Sodium Pyruvate, 50 U/mL Penicillin, 50mg/mL Streptomycin and 50mg/mL Kanamycin
Quantity 75 x 106 Cells