missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Genlantis™ ImmunoPure™ HD Normal Human Peripheral Blood Mononuclear Cells (PBMNCs)
Consists of a variety of immune cells derived from the peripheral blood of normal adults, thereby qualifying them for a diverse number of cell-based experiments
$869.27 - $2014.94
Specifications
Certifications/Compliance | IRB, FDA |
---|---|
Content And Storage | -170°C |
For Use With (Application) | Carry the duty of fighting infections as part of the immune system response |
Media Recommendation | RPMI1640 medium + 10% (v/v) FBS, 2mM Glutamine, 1% (v/v) non-essential Amino Acids, 1% (v/v) Sodium Pyruvate, 50 U/mL Penicillin, 50mg/mL Streptomycin and 50mg/mL Kanamycin |
Product Type | Normal Human Peripheral Blood Mononuclear Cells |
Description
- Proprietary reagents ensure high survival and recovery rates
- Analyzed and characterized by Flow cytometry
- Each lot tested to be free from infectious agents and contaminants
- PBMCs are blood cells with round nucleus and consist of lymphocytes, monocytes, and macrophages
- Lymphocyte and monocyte fractions are isolated from whole blood using apheresis
- These blood cells carry duty of fighting infections as part of immune system response
- PBMNCs are immediately cryopreserved by decreasing temperature by 1°C/minute down to -170°C
- Typically about 75% of the lymphocyte population consists of T cells (CD4+ and CD8+ cells ∽75%), with the other 25% being B cells and Natural Killer (NK) cells
Specifications
IRB, FDA | |
Carry the duty of fighting infections as part of the immune system response | |
Normal Human Peripheral Blood Mononuclear Cells |
-170°C | |
RPMI1640 medium + 10% (v/v) FBS, 2mM Glutamine, 1% (v/v) non-essential Amino Acids, 1% (v/v) Sodium Pyruvate, 50 U/mL Penicillin, 50mg/mL Streptomycin and 50mg/mL Kanamycin | |
Peripheral Blood of Normal Adults |