Please login to your online account to display your discounted pricing
Genlantis™ GenePORTER™ Transfection Reagents and Kits
High-efficiency, serum-free transfection of a broad spectrum of adherent and suspension cells
Specifications
For Use With (Application) Transfection of broad spectrum of adherent and suspension cells, effective DNA complexing, cytoplasmic translocation and subsequent nuclear release of plasmid DNA and DNA Oligonucleotides
Includes Cationic Lipid, Hydration Buffer with Neutral Lipid Dioleoyl Phosphatidylethanolamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    
T201015
gene therapy systems inc
T201015
1.5mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications
Specifications
For Use With (Application) Transfection of broad spectrum of adherent and suspension cells, effective DNA complexing, cytoplasmic translocation and subsequent nuclear release of plasmid DNA and DNA Oligonucleotides
Includes Cationic Lipid, Hydration Buffer with Neutral Lipid Dioleoyl Phosphatidylethanolamine
  • Low cytotoxicity
  • Ideal for delivery of oligonucleotides
  • Unique lipid formulation that allows efficient serum-free transfection of a broad spectrum of adherent and suspension cells
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.