Genlantis™ GenePORTER™-H Transfection Reagent
Hydrated lipid format provides time savings and convenience
Supplier: Genlantis™ T201015H
Description
The reagent consists of the proprietary GenePORTER cationic lipid together with the neutral lipid dioleoyl phosphatidylethanolamine (DOPE) resuspended in a special hydration buffer. These components provide effective DNA complexing, cytoplasmic translocation, and subsequent nuclear release of plasmid DNA and DNA oligonucleotides.
- High-transfection efficiency
- Low cytotoxicity
- Broad-spectrum activity
- Ideal for delivery of oligonucleotides
Specifications
Effective DNA complexing, cytoplasmic translocation and subsequent nuclear release of plasmid DNA and DNA Oligonucleotides | |
Cationic Lipid, Hydration Buffer with Neutral Lipid Dioleoyl Phosphatidylethanolamine | |
1.5 mL |
U.S. Patent 6,348,499