Genlantis™ GenePORTER™-H Transfection Reagent
Hydrated lipid format provides time savings and convenience
$581.12 - $2,454.35
Specifications
For Use With (Application) Effective DNA complexing, cytoplasmic translocation and subsequent nuclear release of plasmid DNA and DNA Oligonucleotides
Includes Cationic Lipid, Hydration Buffer with Neutral Lipid Dioleoyl Phosphatidylethanolamine
View More Specs
Disclaimers:
U.S. Patent 6,348,499
Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T201015H Genlantis
T201015H
1.5mL Each for $581.12
T201075H Genlantis
T201075H
7.5mL Each for $2,454.35
Specifications
Description
The reagent consists of the proprietary GenePORTER cationic lipid together with the neutral lipid dioleoyl phosphatidylethanolamine (DOPE) resuspended in a special hydration buffer. These components provide effective DNA complexing, cytoplasmic translocation, and subsequent nuclear release of plasmid DNA and DNA oligonucleotides.
  • High-transfection efficiency
  • Low cytotoxicity
  • Broad-spectrum activity
  • Ideal for delivery of oligonucleotides
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Effective DNA complexing, cytoplasmic translocation and subsequent nuclear release of plasmid DNA and DNA Oligonucleotides
Cationic Lipid, Hydration Buffer with Neutral Lipid Dioleoyl Phosphatidylethanolamine