Please login to your online account to display your discounted pricing
Genlantis™ GenePORTER™-H Transfection Reagent
Hydrated lipid format provides time savings and convenience
Manufacturer: genlantis  T201075H
 View more versions of this product
 Disclaimers
U.S. Patent 6,348,499
Catalog No. T201075H
  • / Each
Description & Specifications
Specifications
Quantity 7.5mL
For Use With (Application) Effective DNA complexing, cytoplasmic translocation and subsequent nuclear release of plasmid DNA and DNA Oligonucleotides
Includes Cationic Lipid, Hydration Buffer with Neutral Lipid Dioleoyl Phosphatidylethanolamine