Please login to your online account to display your discounted pricing

Genlantis™GenePORTER™ Gold Transfection Reagent

$763.67 - $5,462.98

Specifications

For Use With (Application) Ideal for high throughput applications, for same-day transfection in a broad range of cell lines
Shelf Life 1 Year
Storage Requirements 4°C
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

T204015

 
genlantis
T204015
400 x 1.5mL Each for $763.67

T204030

 
genlantis
T204030
800 x 1.5mL Each for $1,213.65

T204115

 
genlantis
T204115
4000 x 15mL Each for $5,462.98
Description & Specifications

Specifications

For Use With (Application) Ideal for high throughput applications, for same-day transfection in a broad range of cell lines
Shelf Life 1 Year
Storage Requirements 4°C

GenePORTER™ Gold Transfection Reagent is our newest, highly effective and reproducible lipid based gene delivery formulation for the widest variety of cell lines, especially the easy-to-transfect cell lines. It utilizes our Advanced Carrier Enhancement (ACE) technology that maximizes transfection efficiency and transgene expression levels with minimal cytotoxicity. The GenePORTER Gold Reagent and protocol have been optimized to provide the fastest, easiest, and most cost effective transfection reactions possible. Because of its simplicity and cost advantage, the GenePORTER Gold Transfection Reagent is ideal for high throughput or large scale applications where the need for cost control and great results are highly desired.

  • Super fast protocol, optimized for same-day transfection
  • Effective in a broad range of cell lines
  • Ideal for high-throughput applications
  • Value priced - your research dollars will go further
  • Unique Advanced Conjugation Enhancement (ACE) technology promotes effective delivery with exceptionally low cytotoxicity

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

1 Year