Genlantis™ Genlantis™GenePORTER™ Gold Transfection Reagent
Supplier:  Genlantis™ T204030
 View more versions of this product
Catalog No. T204030
Description
Description
GenePORTER™ Gold Transfection Reagent is our newest, highly effective and reproducible lipid based gene delivery formulation for the widest variety of cell lines, especially the easy-to-transfect cell lines. It utilizes our Advanced Carrier Enhancement (ACE) technology that maximizes transfection efficiency and transgene expression levels with minimal cytotoxicity. The GenePORTER Gold Reagent and protocol have been optimized to provide the fastest, easiest, and most cost effective transfection reactions possible. Because of its simplicity and cost advantage, the GenePORTER Gold Transfection Reagent is ideal for high throughput or large scale applications where the need for cost control and great results are highly desired.
  • Super fast protocol, optimized for same-day transfection
  • Effective in a broad range of cell lines
  • Ideal for high-throughput applications
  • Value priced - your research dollars will go further
  • Unique Advanced Conjugation Enhancement (ACE) technology promotes effective delivery with exceptionally low cytotoxicity
Specifications
Specifications
4°C
Ideal for high throughput applications, for same-day transfection in a broad range of cell lines
800 x 1.5 mL