Please login to your online account to display your discounted pricing

Genlantis™ DNA Markers and Ladders


Fragment size from 11 base pairs to 15kb for a variety of electrophoretic applications

Manufacturer: genlantis  D405500

 View more versions of this product

Catalog No. D405500

  • / Each
 

Description & Specifications

Specifications

Shelf Life 1 year
Storage Requirements −20°C
Quantity 500μL
No. of Bands 11
For Use With (Application) Determining the size of DNA fragment
Accessories

1 year