Please login to your online account to display your discounted pricing

Genlantis™ DNA Markers and Ladders


Fragment size from 11 base pairs to 15kb for a variety of electrophoretic applications

Manufacturer: genlantis  D402100

 View more versions of this product

Catalog No. D402100

  • / Each
 

Description & Specifications

Specifications

Storage Requirements −20°C
Shelf Life 1 year
For Use With (Application) Determining the size of DNA fragment
Quantity 100μg
Accessories

1 year