Please login to your online account to display your discounted pricing
Genlantis™ Dicer siRNA Generation Kits
Easily, economically generate functional siRNAs
Manufacturer: genlantis  T510008
 View more versions of this product
Catalog No. T510008
  • / Each
Description & Specifications
Specifications
Includes Recombinant Dicer™ Enzyme kit (0.5U/μL), Dicer™ Reaction Buffer, 50mM MgCl2 solution, 10mM ATP, Dicer™ stop solution, Nuclease-free Water
Quantity 200U
For Use With (Application) Provides sufficient siRNAs in every single species by using ultra-active form of Human recombinant Dicer™ Enzyme to achieve effective gene silencing