Please login to your online account to display your discounted pricing
Genlantis™ Dicer siRNA Generation Kits
Easily, economically generate functional siRNAs
Manufacturer: genlantis  T510003
 View more versions of this product
Catalog No. T510003
  • / Each
Description & Specifications
Specifications
For Use With (Application) Provides sufficient siRNAs in every single species by using ultra-active form of Human recombinant Dicer™ Enzyme to achieve effective gene silencing
Includes T7 Enzyme mix, T7 Reaction Buffer, NTP mix, DNase I , 2X Gel Loading Buffer, Nuclease-free Water, LiCl precipitation solution, GFP control plasmid, 5' control primer, 3' control primer
Quantity 20U