Please login to your online account to display your discounted pricing
Genlantis™ Dicer siRNA Generation Kits
Easily, economically generate functional siRNAs
Manufacturer: genlantis  T510001
 View more versions of this product
Catalog No. T510001
  • / Each
Description & Specifications
Specifications
Includes Recombinant Dicer™ Enzyme kit (50U), TurboScript™ T7 transcription kit (5 rxns), GeneSilencer™ siRNA transfection reagent kit (50 rxns), RNA purification column 1 and 2
For Use With (Application) Provides sufficient siRNAs in every single species by using ultra-active form of Human recombinant Dicer™ Enzyme to achieve effective gene silencing
Quantity 50U